EMAIL THIS PAGE TO A FRIEND
Pharmacology & therapeutics
Antiviral iodinated pyrimidine deoxyribonucleosides: 5-iodo-2'-deoxyuridine; 5-iodo-2'-deoxycytidine; 5-iodo-5'-amino-2',5'-dideoxyuridine.
PMID 392550