EMAIL THIS PAGE TO A FRIEND
[Osaka Daigaku shigaku zasshi] The journal of Osaka University Dental Society
[An approach to caries prevention by a zinc acetate-strontium acetate-tannic acid mixture: biological and physicochemical actions of the mixture on enamel, oral streptococci and dental plaque].
PMID 397328