EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Absolute configuration of 3-hydroxyadipic acid in human urine.
PMID 3980660
Abstract
A method involving derivatization and combined gas chromatography--mass spectrometry has been developed to separate the enantiomers of 3-hydroxyadipic acid. By combining this method with asymmetric synthesis of the same acid, it has been shown that 3-hydroxyadipic acid excreted in urine consists of at least 95% of the L-enantiomer. This finding supports the hypothesis that dicarboxylic acids are degraded by ordinary beta-oxidation, and indicates that adipic acid may be converted into succinic acid.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
92826
Disodium (2RS)-2-hydroxyadipate, ≥95.0% (GC)
C6H8Na2O5