EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
In vitro activities of 47 antimicrobial agents against three Campylobacter spp. from pigs.
PMID 3985597
Abstract
The in vitro activities of 47 antimicrobial agents against 30 isolates of Campylobacter species from pigs were determined by the agar dilution technique. The isolates were obtained from pigs with proliferative enteritis and included 10 strains each of Campylobacter coli, Campylobacter sputorum subsp. mucosalis, and "Campylobacter hyointestinalis Gebhart et al." (this name is not on the Approved Lists). Carbadox, furazolidone, nitrofurantoin, gentamicin, and dimetridazole were the most active drugs, inhibiting all three Campylobacter species with a MIC for 50% of the isolates of 2 micrograms/ml or less. Trimethoprim-sulfamethoxazole, cefazolin, sulfachloropyridazine, novobiocin, vancomycin, sulfathiazole, cyclohexamide, bacitracin, p-arsanilic acid, and colistin were the least active, with MICs for 50% of the isolates ranging from 16 to greater than or equal to 128 micrograms/ml.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D4025
Dimetridazole
C5H7N3O2