EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
An enantioselective synthesis of platelet-activating factors, their enantiomers, and their analogues from D- and L-tartaric acids.
PMID 4017109