EMAIL THIS PAGE TO A FRIEND
FEBS letters
Valosin: isolation and characterization of a novel peptide from porcine intestine.
PMID 4054310
Abstract
The isolation and primary structure of a novel gastrointestinal peptide, designated valosin, is described. The peptide was purified from porcine upper gut extracts using an HPLC and N-terminal sequence screening strategy which depends on chromatographic and structural characteristics as isolation criterion. The amino acid sequence of this peptide consists of 25 amino acid residues:
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V6505
Valosin Porcine, ≥97% (HPLC)
C141H207N31O35S