EMAIL THIS PAGE TO A FRIEND
Cancer research
Growth characteristics and enzyme activities in a survey of transformation markers in adult rat liver epithelial-like cell cultures.
PMID 40692
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
87050701
ARL-6