EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Assignment of three human genes to chromosomes (LDH-A to 11, TK to 17, and IDH to 20) and evidence for translocation between human and mouse chromosomes in somatic cell hybrids (thymidine kinase-lactate dehydrogenase A-isocitrate dehydrogenase-C-11, E-17, and F-20 chromosomes).
PMID 4110482
Abstract
Independently derived man-mouse somatic cell hybrids and their derivative subclones show a positive correlation between the expression of human lactate dehydrogenase A subunits and the occurrence of the human C-11 chromosome. Data are also presented that confirm the previously reported linkage of the thymidine kinase locus to the E-17 chromosome. A translocation of the E-17 chromosome provides presumptive evidence for the assignment of the thymidine kinase locus to the long arm segment of the E-17 chromosome. This translocation also provides evidence for translocation between man and mouse chromosomes in somatic cell hybrids. A presumptive association between the human phenotype for isocitrate dehydrogenase and the human F group is also described. Identification of specific human chromosomes was achieved by the application of several new cytological techniques: measurement of chromosome arm length, in situ annealing with mouse satellite complementary RNA, constitutive heterochromatin staining with Giemsa, and quinacrine mustard fluorochromatic staining.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
90083001
L-M (TK-)