EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Cryoenzymology: the study of enzyme catalysis at subzero temperatures.
PMID 41158
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
50049
Glycine, BioUltra, ≥99.0% (NT)
C2H5NO2