EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Human DNA polymerase 3 (R-DNA polymerase): distinction from DNA polymerase I and reverse transcriptase.
PMID 4129806
Abstract
DNA polymerase III is an enzyme activity in eukaryotic cells which under certain conditions shows strong preference for polyadenylic acid as template when primed by oligodeoxythymidylate. Its first complete separation from other DNA polymerases in human lymphoblasts is reported. This enzyme is biochemically and immunologically distinct from DNA polymerase I and from viral reverse transcriptase from a primtate type C virus.