EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Detection of oxidases on polyacrylamide gels.
PMID 4148790
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N6876
Nitrotetrazolium Blue chloride, ~98% (TLC)
C40H30Cl2N10O6