EMAIL THIS PAGE TO A FRIEND
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society
The unlabeled antibody enzyme method of immunohistochemistry: preparation and properties of soluble antigen-antibody complex (horseradish peroxidase-antihorseradish peroxidase) and its use in identification of spirochetes.
PMID 4192899