EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
N-methylation of biogenic amines. I. Characterization and properties of an N-methyltransferase in rat brain using 5-methyltetrahydrofolic acid as the methyl donor.
PMID 4211218