EMAIL THIS PAGE TO A FRIEND
Journal of cell science
Cytogenetic and nuclear studies on A9 and B82 cells fused together by sendai virus: the early phase.
PMID 4311079
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
85011408
B82