EMAIL THIS PAGE TO A FRIEND
Journal of the National Cancer Institute
Further characterization of the WI-L1 and WI-L2 lymphoblastoid lines.
PMID 4323138
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
90112121
WIL2 NS