EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Adrenocorticotropins. 44. Total synthesis of the human hormone by the solid-phase method.
PMID 4347148
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0423 Adrenocorticotropic Hormone human, ≥97% (HPLC)
C207H308N56O58S