EMAIL THIS PAGE TO A FRIEND
Journal of the National Cancer Institute
Glandular epithelial cells from mice: a method for selective cultivation.
PMID 4363715
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
94081121
NMuMG