EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Purification and properties of halogenated tyrosine and thyroid hormone transaminase from rat kidney mitochondria.
PMID 4381052
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0754
3,5-Diiodo-L-tyrosine dihydrate, crystalline
C9H9I2NO3 · 2H2O