EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
A peptide molecule mimicking the copper(II) transport site of human serum albumin. A comparative study between the synthetic site and albumin.
PMID 4411707