EMAIL THIS PAGE TO A FRIEND
Biochemical medicine
The synthesis and analytical use of a highly sensitive and convenient substrate of elastase.
PMID 4429553
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S4760
N-Succinyl-Ala-Ala-Ala-p-nitroanilide, elastase substrate
C19H25N5O8