EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Structure and tautomerism of the neutral and monoanionic forms of 4-thiouracil derivatives.
PMID 4436501
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
440736
4-Thiouracil, 97%
C4H4N2OS