EMAIL THIS PAGE TO A FRIEND
Pure and applied chemistry. Chimie pure et appliquee
Macrocyclic spermidine and spermine alkaloids.
PMID 4567594
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
49761
Spermidine, analytical standard
C7H19N3
05292
Spermidine 0.1 M solution