EMAIL THIS PAGE TO A FRIEND
Residue reviews
Comment on the distinction between hazard and safety in evaluation of human health hazards on use of dichlorvos, especially in resin strips.
PMID 4570403
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45441
Dichlorvos, PESTANAL®, analytical standard
C4H7Cl2O4P