EMAIL THIS PAGE TO A FRIEND
Life sciences
The roles of the polyamines, putrescine, spermidine, and spermine in normal and malignant tissues.
PMID 4591188
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
51799
Putrescine, analytical standard
C4H12N2
05292
Spermidine 0.1 M solution