EMAIL THIS PAGE TO A FRIEND
The American journal of clinical nutrition
Are nickel, vanadium, silicon, fluorine, and tin essential for man? A review.
PMID 4596029
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF26583194
Tin, foil, 150x150mm, thickness 0.125mm, as rolled, 98.8%
Sn
GF66339254
Tin, foil, 150x150mm, thickness 0.125mm, as rolled, 99.99+%
Sn
GF54760259
Tin, foil, 150x150mm, thickness 0.15mm, as rolled, 98.8%
Sn
GF67327375
Tin, foil, 150x150mm, thickness 0.1mm, as rolled, 98.8%
Sn
GF27720019
Tin, foil, 150x150mm, thickness 0.1mm, as rolled, 99.95%
Sn
GF16685219
Tin, foil, 150x150mm, thickness 0.20mm, as rolled, 98.8%
Sn
GF45567434
Tin, foil, 150x150mm, thickness 0.25mm, as rolled, 99.99+%
Sn
GF28603212
Tin, foil, 150x150mm, thickness 0.5mm, as rolled, 98.8%
Sn
GF28221274
Tin, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99+%
Sn
GF77851323
Tin, foil, 150x150mm, thickness 1.0mm, as rolled, 98.8%
Sn
GF16505636
Tin, foil, 150x150mm, thickness 1.0mm, as rolled, 99.999%
Sn
GF50720925
Tin, foil, 150x150mm, thickness 2.0mm, as rolled, 98.8%
Sn
GF00846413
Tin, foil, 150x150mm, thickness 2.0mm, as rolled, 99.99+%
Sn
GF99151881
Tin, foil, 25x25mm, thickness 0.125mm, as rolled, 98.8%
Sn
GF83193925
Tin, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%
Sn
GF86850029
Tin, foil, 25x25mm, thickness 0.15mm, as rolled, 98.8%
Sn
GF17353425
Tin, foil, 25x25mm, thickness 0.15mm, as rolled, 99.99+%
Sn
GF40941097
Tin, foil, 25x25mm, thickness 0.1mm, as rolled, 98.8%
Sn
GF66628996
Tin, foil, 4mm disks, thickness 0.007mm, 97.4%
Sn
GF33080532
Tin, foil, 4mm disks, thickness 0.015mm, 99.75%
Sn
GF29177039
Tin, foil, 4mm disks, thickness 0.02mm, 99.75%
Sn
GF51371232
Tin, foil, 4mm disks, thickness 0.125mm, as rolled, 98.8%
Sn
GF34214464
Tin, foil, 4mm disks, thickness 0.25mm, as rolled, 98.8%
Sn
GF63204106
Tin, foil, 4mm disks, thickness 1.0mm, as rolled, 100%
Sn
GF04830331
Tin, foil, 50mm disks, thickness 0.0125mm, 97.4%
Sn
GF96864643
Tin, foil, 50mm disks, thickness 0.05mm, as rolled, 98.8%
Sn
GF07913088
Tin, foil, 8mm disks, thickness 0.06mm, as rolled, 99.99+%
Sn
GF18183764
Tin, foil, 8mm disks, thickness 0.1mm, as rolled, 98.8%
Sn
GF36451098
Tin, foil, 8mm disks, thickness 0.65mm, as rolled, 99.99+%
Sn
GF00022699
Tin, foil, light tested, 100x100mm, thickness 0.0125mm, 97.4%
Sn