EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Isolation of protein kinases from reticulocytes and phosphorylation of initiation factors.
PMID 459914
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
NIST975A
Isotopic standard for chlorine, NIST SRM 975a
ClNa
746398
Sodium chloride, anhydrous, Redi-Dri, free-flowing, ACS reagent, ≥99%
ClNa
RES0926S-A7
Sodium chloride, PharmaGrade, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
ClNa
793566
Sodium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
ClNa
S7653
Sodium chloride, BioXtra, ≥99.5% (AT)
ClNa
S3014
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥98% (titration)
ClNa
S5886
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
ClNa
S1679
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
ClNa
S6150
Sodium chloride, tablet
ClNa
S6191
Sodium chloride, BioPerformance Certified, ≥99% (titration), Cell Culture Tested
ClNa
450006
Sodium chloride, anhydrous, beads, −10 mesh, 99.999% trace metals basis
ClNa
450014
Sodium chloride, anhydrous, beads, −10 mesh, 99.99% trace metals basis
ClNa
204439
Sodium chloride, 99.999% trace metals basis
ClNa
378860
Sodium chloride, random crystals, optical grade, 99.98% trace metals basis
ClNa
S9888
Sodium chloride, ACS reagent, ≥99.0%
ClNa
S9625
Sodium chloride, ReagentPlus®, ≥99%
ClNa
310166
Sodium chloride, reagent grade, ≥98%, +80 mesh particle size
ClNa
71376
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
ClNa
71382
Sodium chloride, puriss. p.a., ≥99.5% (AT), powder or crystals
ClNa
71380
Sodium chloride, puriss. p.a., ≥99.5% (AT)
ClNa
31434
Sodium chloride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5%
ClNa
13423
Sodium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5% (calc. to the dried substance), ≤0.00002% Al
ClNa
1613804
Sodium chloride, United States Pharmacopeia (USP) Reference Standard
ClNa
NIST2201
Sodium chloride, NIST® SRM® 2201, ion-selective
ClNa
NIST919B
Sodium chloride, NIST® SRM® 919b
ClNa
71383
Sodium chloride, tested according to Ph.Eur.
ClNa
71387
Sodium chloride, certified reference material for titrimetry, certified by BAM, according to ISO 17025, ≥99.5%
ClNa
28-2270
Sodium chloride, JIS special grade, ≥99.5%
ClNa
PHR1321
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
ClNa