EMAIL THIS PAGE TO A FRIEND
In vitro
Clonal isolation of differentiated rat lung cells.
PMID 4616002
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
90112811
L-2