EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Microbial oxidation of 4-chloroaniline.
PMID 4682327
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
477222
4-Chloroaniline, purified by sublimation, ≥99%
C6H6ClN