EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Tautomerism of the nucleoside antibiotic formycin, as studied by carbon-13 nuclear magnetic resonance.
PMID 4730666
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00287
Formycin A, from Streptomyces kaniharaensis, ≥98% (HPLC)
C10H13N5O4