EMAIL THIS PAGE TO A FRIEND
Journal of cell science
The analysis of malignancy by cell fusion. IV. Hybrid between tumour cells and a malignant L cell derivative.
PMID 4734738
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
88042804
A9HT