EMAIL THIS PAGE TO A FRIEND
Archiv fur klinische und experimentelle Dermatologie
[Present status of griseofulvin therapy of tinea pedis].
PMID 4862059
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G0600000
Griseofulvin, European Pharmacopoeia (EP) Reference Standard
C17H17ClO6