EMAIL THIS PAGE TO A FRIEND
Journal of ultrastructure research
Rapid and improved methods fr embedding biological tissues in Epon 812 and Araldite 502.
PMID 4865651
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3415
Bisphenol A diglycidyl ether
C21H24O4