EMAIL THIS PAGE TO A FRIEND
Endocrinology
Studies on interstitial cells in tissue culture. II. Steroid biosynthesis by a clonal line of rat testicular interstitial cells.
PMID 4868022
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
89031606
R2C