EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
The chemical structure of tryptophanase from Escherichia coli. I. Isolation and structure of a pyridoxyl decapeptide from borohydride-reduced holotryptophanase.
PMID 4912534