EMAIL THIS PAGE TO A FRIEND
Journal of molecular biology
Molecular weights of coliphages and coliphage DNA. IV. Molecular weights of DNA from bacteriophages T4, T5 and T7 and the general problem of determination of M.
PMID 4923669