EMAIL THIS PAGE TO A FRIEND
The British journal of dermatology
Therapeutics. XIV. Herpes simplex virus infections and idoxuridine.
PMID 4932957
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I0050000
Idoxuridine, European Pharmacopoeia (EP) Reference Standard
C9H11IN2O5