EMAIL THIS PAGE TO A FRIEND
Annals of the New York Academy of Sciences
An orphan virus isolated in marine fish cell tissue culture.
PMID 4957055
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
88010601
Grunt Fin (GF)