EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Biological activity and enzymic degradation of substance P analogs: implications for studies of the substance P receptor.
PMID 496986