EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Purification of dipeptidyl aminopeptidase II (dipeptidyl arylamidase II) of the anterior pituitary gland. Peptidase and dipeptide esterase activities.
PMID 4969969