EMAIL THIS PAGE TO A FRIEND
Biochemistry
Inhibition of intractable nucleases with ribonucleoside--vanadyl complexes: isolation of messenger ribonucleic acid from resting lymphocytes.
PMID 497174
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R3380 Ribonucleoside vanadyl complexes, 200 mM