EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
D-Fucose metabolism in a pseudomonad. IV. Cleavage of 2-keto-3-deoxy-D-fuconate to pyruvate and D-lactaldehyde by 2-keto-3-deoxy-L-arabonate aldolase.
PMID 5016652
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
42418
D-Lactaldehyde solution, 1 M in H2O
C3H6O2