EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
A total synthesis of ( )-fumagillin.
PMID 5016935
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
249904
3,3-Dimethylallyl bromide, technical grade, ~90%
C5H9Br