EMAIL THIS PAGE TO A FRIEND
Medical laboratory technology
Arizona infection in turkeys in Great Britain.
PMID 5104952
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
62915 Lysine Iron Agar, for microbiology