EMAIL THIS PAGE TO A FRIEND
Molecular pharmacology
The use of a cholinergic fluorescent probe for the study of the receptor proteolipid.
PMID 5139564
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D2762
(Dansylaminoethyl)­tri­methyl­ammonium perchlorate
C17H26N3O2S · ClO4