EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Affinity chromatography of thrombin.
PMID 5168884
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9527
ε-Aminocaproyl-p-chlorobenzylamide hydrochloride
C13H19ClN2O · HCl