EMAIL THIS PAGE TO A FRIEND
Japanese journal of pharmacology
Newer pharmacological data on oxyphenbutazone.
PMID 5320527
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0540 Oxyphenbutazone, ≥98% (HPLC)
C19H20N2O3