EMAIL THIS PAGE TO A FRIEND
Advances in tracer methodology
Diisopropylfluorophosphate-P32 (DFP-P32) as a label for granulocytes.
PMID 5334301
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0879
Diisopropylfluorophosphate
C6H14FO3P