EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Analysis of free phytosterols by gas chromatography using liquid phase OV-101.
PMID 5344164
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
21105 GC Stationary Phase, phase OV-17, bottle of 25 g