EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical sciences
Rapid peptide synthesis: synthesis of human fibrinopeptide A.
PMID 5353289
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F3254
Fibrinopeptide A human, ≥96% (HPLC)
C63H97N19O26