EMAIL THIS PAGE TO A FRIEND
European journal of biochemistry
Yeast alcohol dehydrogenase: SH groups, disulfide groups, quaternary structure, and reactivation by reductive cleavage of disulfide groups.
PMID 5353605